missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) chloride heptahydrate, 99%

Product Code. 11499783
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499783 1000 g 1000g
11409793 50 g 50g
11419793 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499783 Supplier Thermo Scientific Alfa Aesar Supplier No. A12947.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18618-55-8 | CeCl3·7H2O

Cerium(III) chloride heptahydrate is used in the preparation of allylsilanes from esters. It is used as a reducing agent in organic synthesis in place of sodium borohydride. In Luche reaction, carvone gives selectively allylic alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(III) chloride heptahydrate is used in the preparation of allylsilanes from esters. It is used as a reducing agent in organic synthesis in place of sodium borohydride. In Luche reaction, carvone gives selectively allylic alcohol.

Solubility
Soluble in alcohol and acetone. Slightly soluble in tetrahydrofuran. Insoluble in cold water.

Notes
Air sensitive. Hygroscopic. Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18618-55-8
Molecular Formula CeCl3·7H2O
MDL Number MFCD00149634
Synonym Cerous chloride heptahydrate

Specifications

Quantity 1000 g
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,1997
Solubility Information Soluble in alcohol,water,and acetone. Slightly soluble in tetrahydrofuran.
Formula Weight 372.60 (246.48 Anhydrous)
Percent Purity 99%
Chemical Name or Material Cerium(III) chloride heptahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.