missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) Chloride Heptahydrate, TRC

Product Code. 30359320
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30359320 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30359320

Brand: Toronto Research Chemicals TRCC2770585G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cerium(III) Chloride Heptahydrate
CAS 18618-55-8
Molecular Formula CeCl3 . 7(H2O)
Quantity 5 g
InChI Formula InChI=1S/Ce.3ClH.7H2O/h;3*1H;7*1H2/q+3;;;;;;;;;;/p-3
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Ce+3]
IUPAC Name cerium(3+);trichloride;heptahydrate
Molecular Weight (g/mol) 372.58
Formula Weight 370.8859
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.