missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) trifluoromethanesulfonate, 98%

Product Code. 11359106
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359106 5 g 5g
11369106 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359106 Supplier Thermo Scientific Alfa Aesar Supplier No. L20251.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76089-77-5 | C3CeF9O9S3 | 587.31 g/mol

It is employed as intermediate for organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for organic synthesis and pharmaceuticals.

Solubility
Soluble in water.

Notes
Hygroscopic. store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76089-77-5
Molecular Formula C3CeF9O9S3
Molecular Weight (g/mol) 587.31
MDL Number MFCD02093899
InChI Key PHSMPGGNMIPKTH-UHFFFAOYSA-K
Synonym cerium iii trifluoromethanesulfonate, cerium iii triflate, trifluoromethanesulfonic acid cerium iii salt, cerium trifluoromethanesulfonate, cerous triflate, ce otf 3, 3cf3o3s.ce, acmc-209p1w, ksc491c9f
PubChem CID 2733941
IUPAC Name cerium(3+);trifluoromethanesulfonate
SMILES [Ce+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point >300°C
Density 1.7
Linear Formula (CF3SO3)3Ce
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 587.33
Percent Purity 98%
Chemical Name or Material Cerium(III) trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.