missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol

Product Code. 11398438
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11398438

Brand: Thermo Scientific Alfa Aesar 040471.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 876107-33-4 | C12H28CeO8 | 440.464 g/mol

Hydrolysis of cerium 2-methoxyethoxide and subsequent supercritical drying or freeze-drying yielded a CeO 2 aerogel or cryogel with quite a large surface area above 300 m 2 /g.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 876107-33-4
Molecular Formula C12H28CeO8
Molecular Weight (g/mol) 440.464
MDL Number MFCD00236530
InChI Key JRRALIVAHBLZSS-UHFFFAOYSA-N
Synonym cerium iv methoxyethoxide w/w in methoxyethanol, tetrakis 2-methoxyethoxy cerium iv, cerium 4+ tetrakis 2-methoxyethan-1-olate, cerium iv methoxyethoxide in methoxyethanol
PubChem CID 21200429
IUPAC Name cerium(4+);2-methoxyethanolate
SMILES COCC[O-].COCC[O-].COCC[O-].COCC[O-].[Ce+4]

Specifications

Color Yellow
Density 1.02 g/mL
Linear Formula Ce[OCH2CH2OCH3]4
Quantity 25 g
UN Number UN1188
Sensitivity Moisture sensitive
Solubility Information Reacts with air.
Formula Weight 440.47
Concentration or Composition (by Analyte or Components) 18 to 20% w/w in 2-methoxyethanol
Physical Form Liquid
Chemical Name or Material Cerium(IV) 2-methoxyethoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.