missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) ammonium nitrate, 98+%

Product Code. 11455323
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455323 250 g 250g
11445323 50 g 50g
11435323 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455323 Supplier Thermo Scientific Alfa Aesar Supplier No. A12882.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Cerium(IV) ammonium nitrate, >-98%, H8CeN8O18, CAS Number-16774-21-3, Ammonium cerium(IV) nitrate; CAN, 1000g, Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S., 240-827-6, 548.23, H272-H290-H302-H314-H317-H335 | CAS: 16774-21-3 | H8CeN8O18

Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.

Solubility
Soluble in water and alcohol.

Notes
Store in cool place. Hygroscopic. Incompatible with strong reducing agents and powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16774-21-3
Molecular Formula H8CeN8O18
MDL Number MFCD00151121
Synonym Ammonium cerium(IV) nitrate; CAN

Specifications

Quantity 250 g
Linear Formula (NH4)2Ce(NO3)6
UN Number UN1477
Merck Index 14,1992
Solubility Information Soluble in water and alcohol.
Formula Weight 548.23
Percent Purity ≥98%
Odor Pungent
Chemical Name or Material Cerium(IV) ammonium nitrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.