Learn More
Cerium(IV) isopropoxide, Ce 37-45%
Description
Cerium(IV) isopropoxide, Ce 37-45% is used to catalyzes diastereoselective pinacol couplings. It forms diketonates ,which hydrolyze to CeO2 particles. CeO2 particles have the potential for ion conduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 140°C (decomposition) |
| Color | Orange-Red to Yellow |
| Linear Formula | Ce(OC3H7)4 |
| Quantity | 5 g |
| UN Number | UN2925 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in pyridine |
| Formula Weight | 376.48 |
| Concentration or Composition (by Analyte or Components) | Ce 37 to 45% |
| Physical Form | Powder |
| Show More |
Safety and Handling
- Cerium(IV) isopropoxide
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 C
- H228-Flammable solid.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Cerium(IV) isopropoxide
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.