missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) oxide, 99.9%, (trace metal basis)

Product Code. 10571451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10571451 500 g Plastic bottle 500g
10470281 100 g Plastic bottle 100g
13438448 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10571451 Supplier Thermo Scientific Acros Supplier No. 199125000
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1306-38-3 | CeO2 | 172.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-38-3
Molecular Formula CeO2
Molecular Weight (g/mol) 172.12
MDL Number MFCD00010933
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
Synonym ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
PubChem CID 73963
ChEBI CHEBI:79089
IUPAC Name dioxocerium
SMILES O=[Ce]=O

Specifications

Melting Point 2600.0°C
Color White to Yellow
Physical Form Powder
Quantity 500 g
Packaging Plastic bottle
Specific Gravity 7.13
Assay Percent Range 99.9%
Density 7.1300g/mL
Linear Formula CeO2
Fieser 06,99
Merck Index 15,1988
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
Formula Weight 172.12
Percent Purity 99.9%
Grade Trace Metal Basis
Chemical Name or Material Cerium(IV) oxide, 99.9%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.