missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) Oxide, Extra Pure, SLR, Low In Other Rare Earths, Fisher Chemical

Product Code. 10478780 Shop All Fisher Chemical Products
Change view
Click to view available options
100g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10478780 100g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10478780 Supplier Fisher Chemical Supplier No. C/3680/48
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 2600°C
CAS 1306-38-3
Color Yellow
Physical Form Powder/Solid
Quantity 100 g
Molecular Formula CeO2
MDL Number 10933
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
SMILES O=[Ce]=O
IUPAC Name dioxocerium
Molecular Weight (g/mol) 172.11
Formula Weight 172.12g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.