missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate, 95%

Product Code. 15304988 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15304988 100 g 100g
15375137 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15304988 Supplier Thermo Scientific Acros Supplier No. 404341000
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13590-82-4 | CeO8S2 | 332.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13590-82-4
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
Synonym ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
PubChem CID 159684
IUPAC Name cerium(4+);disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 195.0°C
Quantity 100 g
Assay Percent Range 39.6 to 44.7% (Ce) (Iodometry)
Linear Formula Ce(SO4)2
Merck Index 15,1989
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dilute acids
Formula Weight 332.23
Percent Purity 95%
Chemical Name or Material Cerium(IV) sulfate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cerium(IV) sulfate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulfuric acid, cerium(4+) salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.