missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(IV) Sulfate, Concentrated Solution 0.05M, (0.05N), Standard Concentrate, for volumetric analysis, Fisher Chemical
Shop All Fisher Chemical ProductsSpecifications
Specifications
| CAS | 13590-82-4 |
| Physical Form | Liquid |
| Quantity | 6 AMP |
| Molecular Formula | CeO8S2 |
| MDL Number | MFCD00148852 |
| Synonym | ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate |
| InChI Key | VZDYWEUILIUIDF-UHFFFAOYSA-J |
| SMILES | [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| IUPAC Name | λ⁴-cerium(4+) disulfate |
| Molecular Weight (g/mol) | 332.23 |
| Show More |
Safety and Handling
Product Identifier
- Cerium (IV) sulfate, 0.05M, concentrated volumetric solution
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P310-Immediately call a POISON CENTER/doctor/.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- MIXTURE LIST-Contain: Sulfuric acid, cerium(4+) salt (2:1), Sulfuric acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?