missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate solution, 0.05M Ce(SO4)2, Solstice

Product Code. 15614130
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15614130 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15614130 Supplier Honeywell-Fluka Supplier No. 342531L
  • ONLINE EXCLUSIVE PRICE
    163.00 €
    147.33 € / 1 L
    Save 15.67 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 13590-82-4
Quantity 1 L
Molecular Formula CeO8S2
Linear Formula Ce(SO4)2
MDL Number MFCD00148852
UN Number NONH for all modes of transport
Synonym ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name cerium(4+);disulfate
Molecular Weight (g/mol) 332.23
PubChem CID 159684
Formula Weight 332.24g/mol
Concentration or Composition (by Analyte or Components) 0.05M Ce(SO4)2
Chemical Name or Material Cerium(IV) sulfate solution
Show More Show Less
compliance-icons
Product Identifier
  • Cerium(IV) sulfate solution
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P234-Keep only in original container.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.