missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate solution, 0.1M Ce(SO4)2, Solstice

Product Code. 15694780 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15694780 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15694780 Supplier Honeywell-Fluka Supplier No. 350661L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Volumetric, 0.1 M Ce(SO4)2 (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 13590-82-4
Quantity 1 L
Molecular Formula CeO8S2
Linear Formula Ce(SO4)2
MDL Number MFCD00148852
UN Number UN2796
Synonym ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name λ⁴-cerium(4+) disulfate
Molecular Weight (g/mol) 332.23
PubChem CID 159684
Formula Weight 332.24g/mol
Concentration or Composition (by Analyte or Components) 0.1M Ce(SO4)2
Chemical Name or Material Cerium(IV) sulfate solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.