missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate solution 0.1, (0.1N), NIST standard solution ready to use, for volumetric analysis, Fisher Chemical™

Product Code. 10244340 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Amber glass bottle
Bag in box
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10244340 1 L Amber glass bottle 1 L
10635302 2.5 L Amber glass bottle 2.5 L
11492704 10 L Bag in box 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10244340 Supplier Fisher Chemical Supplier No. J/3000/15
  • ONLINE EXCLUSIVE PRICE
    131.08 €
    92.10 € / 1 L
    Save 38.98 €
    29% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 13590-82-4
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
Synonym ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
PubChem CID 159684
IUPAC Name cerium(4+);disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Yellow
Physical Form Liquid
Quantity 1 L
Packaging Amber glass bottle
Vapor Pressure 14mmHg at 20°C
Density Vapor Density: 0.7
compliance-icons
Product Identifier
  • Cerium (IV) sulfate 0.1M in ca. 10% w/v sulfuric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Sulfuric acid, Sulfuric acid, cerium(4+)salt (2:1)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.