missing translation for 'onlineSavingsMsg'
Learn More

Cerium(Iv) Sulfate Tetrahydrate, Honeywell Fluka™

Product Code. 15673470 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
500 g
Unit Size:
25g
500g
Product Code. Quantity unitSize
15673470 500 g 500g
15663470 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 15673470

Brand: Honeywell-Fluka 31606500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10294-42-5
Molecular Formula CeH8O12S2
Molecular Weight (g/mol) 404.288
MDL Number MFCD00148852
InChI Key MEXSQFDSPVYJOM-UHFFFAOYSA-J
Synonym cerium iv sulfate tetrahydrate, ceric sulfate tetrahydrate, unii-6sax34b8je, 6sax34b8je, acmc-20ecws, ceric sulphate tetrahydrate, ksc493s0d, aronis24302, sulfuric acid, cerium 4+ salt 2:1 , tetrahydrate, cerium iv sulfate hydrate
PubChem CID 22723557
IUPAC Name cerium(4+);disulfate;tetrahydrate
SMILES O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]

Specifications

Quantity 500 g
Linear Formula Ce(SO4)2 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 404.3g/mol
Percent Purity ≥98%
Grade puriss. p.a.
Chemical Name or Material Cerium(Iv) Sulfate Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.