missing translation for 'onlineSavingsMsg'
Learn More

Cerium rod, 12.7mm (0.5in) dia, 99.8% (metals basis excluding Ta)

Product Code. 11373798
Change view
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
Unit Size:
100mm
10mm
25mm
50mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373798 25 mm 25mm
11393798 10 mm 10mm
11383798 50 mm 50mm
11303808 100 mm 100mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11373798 Supplier Thermo Scientific Alfa Aesar Supplier No. 040284.DH
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7440-45-1 | Ce | 140.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-45-1
Molecular Formula Ce
Molecular Weight (g/mol) 140.12
MDL Number MFCD00010924
InChI Key GWXLDORMOJMVQZ-UHFFFAOYSA-N
Synonym cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
PubChem CID 23974
ChEBI CHEBI:33369
IUPAC Name cerium
SMILES [Ce]

Specifications

Color Silver
Quantity 25 mm
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.86g/mm
Form Cast Rod
UN Number UN1333
Percent Purity 99.9%
Assay (metals basis excluding Ta)
Chemical Name or Material Cerium rod, 12.7mm (0.5 in.) dia.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.