missing translation for 'onlineSavingsMsg'
Learn More

Cesium acetate, 99.998% (metals basis)

Product Code. 11470730
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470730 100 g 100g
11460730 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470730 Supplier Thermo Scientific Alfa Aesar Supplier No. 044434.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3396-11-0 | C2H3CsO2 | 191.949 g/mol

Cesium acetate is used in organic synthesis especially in the Perkin reaction to prepare unsaturated cinnamic-type acids. It is used in petroleum drilling fluids as an alternative to cesium formate. In organometallic chemistry, it is an additive for metal-ctalyzed coupling reactions. It is also used in the inversion of stereo chemistry of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3396-11-0
Molecular Formula C2H3CsO2
Molecular Weight (g/mol) 191.949
MDL Number MFCD00013056
InChI Key ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
Synonym cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
PubChem CID 5152919
IUPAC Name cesium;acetate
SMILES CC(=O)[O-].[Cs+]

Specifications

Melting Point 194°C
Physical Form Lump/Powder
Quantity 100 g
Solubility Information Soluble in water and dimethyl sulfoxide.
Formula Weight 191.95
Percent Purity 99.998%
Odor Weak
Assay (metals basis)
Chemical Name or Material Cesium acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.