missing translation for 'onlineSavingsMsg'
Learn More

Cesium bromide, 99.9%, pure

Product Code. 10437780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10437780 25 g Glass bottle 25g
10447780 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10437780 Supplier Thermo Scientific Acros Supplier No. 192060250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7787-69-1 | BrCs | 212.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-69-1
Molecular Formula BrCs
Molecular Weight (g/mol) 212.81
MDL Number MFCD00010954
InChI Key LYQFWZFBNBDLEO-UHFFFAOYSA-M
Synonym cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide
PubChem CID 24592
IUPAC Name cesium;bromide
SMILES [Br-].[Cs+]

Specifications

Boiling Point 1300.0°C
Melting Point 636.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Packaging Glass bottle
Specific Gravity 4.44
Assay Percent Range 99.90%
Density 4.4400g/mL
Linear Formula CsBr
Merck Index 15, 2008
Solubility Information Solubility in water: 55g/100mL (20 c). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
Formula Weight 212.81
Percent Purity 99.9%
Grade Pure
Chemical Name or Material Cesium bromide, 99.9%
Show More Show Less
compliance-icons
Product Identifier
  • Cesium bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.