missing translation for 'onlineSavingsMsg'
Learn More

Cesium bromide, 99.9%, pure

Product Code. p-222816 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10437780

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7787-69-1 | BrCs | 212.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-69-1
Molecular Formula BrCs
Molecular Weight (g/mol) 212.81
MDL Number MFCD00010954
InChI Key LYQFWZFBNBDLEO-UHFFFAOYSA-M
Synonym cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide
PubChem CID 24592
IUPAC Name cesium;bromide
SMILES [Br-].[Cs+]

Specifications

Boiling Point 1300.0°C
Melting Point 636.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 99.90%
Linear Formula CsBr
Merck Index 15, 2008
Solubility Information Solubility in water: 55g/100mL (20 c). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
Formula Weight 212.81
Percent Purity 99.9%
Grade Pure
Packaging Glass bottle
Specific Gravity 4.44
Density 4.4400g/mL
Chemical Name or Material Cesium bromide, 99.9%
Show More Show Less
compliance-icons
Product Identifier
  • Cesium bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.