missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, 99.999% (Metals Basis), Molecular Biology Grade, Ultrapure

Product Code. 15825228 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 15825228

Brand: Thermo Scientific J75822.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for density gradient high speed centrifugation separation of λ-phage, M13 and plasmid DNA. | CAS: 7647-17-8 | ClCs | 168.36 g/mol

Used for density gradient high speed centrifugation separation of λ phage, M13 and plasmid DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1303°C
Melting Point 646°C
Physical Form Crystalline Powder
Quantity 500 g
IUPAC Name cesium;chloride
Formula Weight 168.36
Percent Purity 99.999%
Grade Molecular Biology
Packaging Plastic bottle
Specific Gravity 3.99
Density 3.99
Assay Metal basis
Chemical Name or Material Cesium Chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.