missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, 99.999%, (trace metal basis), extra pure

Product Code. 11924191 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11924191

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-17-8 | ClCs | 168.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290.0°C
Melting Point 646.0°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.999%
Linear Formula CsCl
Merck Index 15,201
Solubility Information Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol, insoluble in acetone
IUPAC Name cesium;chloride
Formula Weight 168.36
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Cesium chloride
Show More Show Less
compliance-icons
Product Identifier
  • Cesium chloride
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.