missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Specifications
Specifications
| Chemical Name or Material | Cesium chloride Lanthanum chloride buffer |
| Quantity | 250 mL |
| MDL Number | MFCD00152971 |
| UN Number | UN3082 |
| Packaging | Poly Bottle |
| Density | 1.17g/mL at 20°C |
Safety and Handling
Product Identifier
- Cesium chloride Lanthanum chloride buffer
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 2
- Skin sensitiser Category 1
- H290-May be corrosive to metals.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H361-Suspected of damaging fertility or the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P234-Keep only in original container.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: Caesium chloride, Lanthanum chloride anhydrous
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?