missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, TraceSELECT™, ≥99.99% (Metals Basis), Honeywell Fluka™

Product Code. 15617410 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15617410

Brand: Honeywell-Fluka 9003325G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TraceSELECT™, for trace analysis,≥99.9995% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]

Specifications

Melting Point 642°C
Color Colorless
Quantity 25 g
Linear Formula CsCl
UN Number NONH for all modes of transport
Solubility Information In water 1000g/L at 20°C
Formula Weight 168.36g/mol
Percent Purity ≥99.9995% (metals basis)
Grade TraceSELECT
Odor Odorless
Packaging Poly Bottle
Appearance Powder
Density 3.98g/cm3 at 20°C
Chemical Name or Material Cesium chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.