missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- For Density Gradient Centrifugation; For Laboratory,Research,or Manufacturing Use
Specifications
Specifications
| Quantity | 100 g |
| Formula Weight | 168.36 |
| Grade | Ultrapure Bioreagent |
| Packaging | Poly bottle |
| Specific Gravity | 1L = 3.99kg |
| Chemical Name or Material | Cesium Chloride |
Safety and Handling
Product Identifier
- Cesium chloride
- Warning
- Reproductive toxicity Category 2
- H361-Suspected of damaging fertility or the unborn child.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Cesium chloride
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?