missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents

Product Code. 10629553
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10629553 1 kg Poly Bottle 1kg
10734344 100 g Poly Bottle 100g
10629743 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10629553 Supplier Fisher BioReagents Supplier No. BP2101
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This Molecular Biology-grade cesium chloride is ideal for UV analysis of nucleic acids during density-gradient centrifugation and for the isolation of plasmid DNA, phage DNA, virus particles, and RNA.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290°C
Melting Point 646°C
Color White
pH 5 to 9
Physical Form Crystalline Powder
Quantity 1 kg
Packaging Poly Bottle
Refractive Index 1.367 to 1.371 (at 25°C)
Assay Percent Range ≥99.999 %
Merck Index 15, 2010
Formula Weight 168.36
Percent Purity ≥99.999%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Cesium Chloride
Show More Show Less
compliance-icons
Product Identifier
  • Cesium chloride
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.