missing translation for 'onlineSavingsMsg'
Learn More

Cesium hydrogen carbonate, 99.99% (metals basis)

Product Code. 11307807
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307807 25 g 25g
11397797 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307807 Supplier Thermo Scientific Alfa Aesar Supplier No. 014234.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29703-01-3 | CHCsO3 | 193.92 g/mol

Cesium hydrogen carbonate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29703-01-3
Molecular Formula CHCsO3
Molecular Weight (g/mol) 193.92
MDL Number MFCD00010961
InChI Key ZMCUDHNSHCRDBT-UHFFFAOYSA-M
Synonym cesium bicarbonate, cesium hydrogen carbonate, caesium bicarbonate, caesium 1+ ion bicarbonate, cesium hydrogencarbonate, acmc-1cmhe, cho3.cs, caesium 1+ bicarbonate, carbonic acid, cesiumsalt 1:1
PubChem CID 2734983
IUPAC Name caesium(1+) hydrogen carbonate
SMILES [Cs+].OC([O-])=O

Specifications

Melting Point 175°C (decomposition)
Physical Form Crystalline Aggregates
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 193.92
Percent Purity 99.99%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium hydrogen carbonate
compliance-icons compliance-icons
Product Identifier
  • Cesium hydrogen carbonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Carbonic acid, cesium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.