missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Hydroxide Monohydrate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Cesium Hydroxide Monohydrate |
| CAS | 35103-79-8 |
| Molecular Formula | CsHO . H2O |
| Quantity | 5 g |
| InChI Formula | InChI=1S/Cs.2H2O/h;2*1H2/q+1;;/p-1 |
| SMILES | O.[OH-].[Cs+] |
| IUPAC Name | cesium;hydroxide;hydrate |
| Molecular Weight (g/mol) | 167.928 |
| Formula Weight | 167.9188 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction