missing translation for 'onlineSavingsMsg'
Learn More

Cesium iodide, ultra dry, 99.998% (metals basis)

Product Code. 11368597
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
Unit Size:
10g
1g
250g
50g
This item is not returnable. View return policy

Product Code. 11368597

Brand: Thermo Scientific Alfa Aesar 035729.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-17-5 | CsI | 259.81 g/mol

Cesium iodide is used as a beam splitter in Fourier transform infrared (FT-IR) spectrometers. It is widely used as a scintillator in silicon photodiodes. It is utilized in calorimeters and detectors and also employed as phosphor in fluoroscopy equipments. It finds application in making cell windows and prisms in infrared spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 7789-17-5
Molecular Formula CsI
Molecular Weight (g/mol) 259.81
MDL Number MFCD00010962
InChI Key XQPRBTXUXXVTKB-UHFFFAOYSA-M
Synonym cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
PubChem CID 24601
SMILES [I-].[Cs+]

Specifikationer

Boiling Point 1280°C
Melting Point 626°C
Color White
Physical Form Beads
Quantity 50 g
Merck Index 14,2014
Solubility Information Soluble in water and ethanol. Slightly soluble in methanol. Insoluble in acetone.
IUPAC Name cesium;iodide
Formula Weight 259.81
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Refractive Index 1.7876
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium iodide, Ultra dry
Visa mer Visa mindre

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.