missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Cetirizine-d4 (ethoxy-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (±)-Cetirizine-d4 (ethoxy-d4) |
| CAS | 1219803-84-5 |
| Molecular Formula | C21 D4 H21 Cl N2 O3 |
| Quantity | 0.01 g |
| Synonym | 2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]-1,1,2,2-tetradeuterioethoxy]acetic acid, 2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy-1,1,2,2-d4]acetic acid, Cetirizine-d4, Cetirizine-D4 (ethoxy-1,1,2,2-D4), (±)-Cetirizine-d4 (ethoxy-d4) |
| InChI Formula | InChI=1 S/C21H25ClN2O3/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)24-12-10-23(11-13-24)14-15-27-16-20(25)26/h1-9,21 H,10-16H2,(H,25,26)/i14D2,15D2 |
| SMILES | [2 H]C([2 H])(OCC(=O)O)C([2 H])([2 H])N1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| IUPAC Name | 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]-1,1,2,2-tetradeuterioethoxy]acetic acid |
| Molecular Weight (g/mol) | 392.9124 |
| Formula Weight | 392.1805 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction