missing translation for 'onlineSavingsMsg'
Learn More

Cetyl Alcohol, Solstice

Product Code. 15660700
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15660700 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15660700 Supplier Honeywell-Riedel-de Haen Supplier No. 03884500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tested according to Ph.Eur.

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 36653-82-4
Melting Point 49°C to 50°C
Boiling Point 190°C
Flash Point 170°C (338°F)
Packaging Poly bottle
Molecular Formula C16H34O
Linear Formula CH3(CH2)15OH
MDL Number MFCD00004760
Quantity 500 g
UN Number NONH for all modes of transport
Beilstein 1748475
Synonym 1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
InChI Key BXWNKGSJHAJOGX-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCO
IUPAC Name hexadecan-1-ol
Molecular Weight (g/mol) 242.45
PubChem CID 2682
ChEBI CHEBI:16125
Formula Weight 242.44g/mol
Chemical Name or Material Cetyl Alcohol, Tested According to Ph.Eur.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.