missing translation for 'onlineSavingsMsg'
Learn More

Cetyltrimethylammonium bromide, ∽98%, Molecular biology reagent grade, MP Biomedicals™

Product Code. 11466461 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Unit Size:
100g
1kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466461 100 g 100g
11476461 250 g 250g
11486461 500 g 500g
11496461 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11466461 Supplier MP Biomedicals Supplier No. 0219400480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemische Identifikatoren

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
Synonym cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon
PubChem CID 5974
ChEBI CHEBI:3567
IUPAC Name hexadecyl(trimethyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Spezifikation

Color White
Quantity 100 g
Formula Weight 364.5
Percent Purity ∼98%
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material Cetyltrimethylammonium bromide
compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Cetyltrimethylammonium Bromide, molecular biology reagent
Signalwort
  • Danger
Gefahrenkategorie
  • Specific target organ toxicity after repeated exposure Category 2
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
Sicherheitshinweis
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.