missing translation for 'onlineSavingsMsg'
Learn More

Cetyltrimethylammonium Bromide, MP Biomedicals

Product Code. 11478094 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478094 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11478094 Supplier MP Biomedicals Supplier No. 0520823280
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 57-09-0
Quantity 100 g
Molecular Formula C19H42BrN
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
IUPAC Name hexadecyltrimethylazanium bromide
Molecular Weight (g/mol) 364.46
PubChem CID 5974
ChEBI CHEBI:3567
Formula Weight 364.46
Chemical Name or Material Cetyltrimethylammonium Bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.