missing translation for 'onlineSavingsMsg'
Learn More

CF 1743, TRC

Product Code. 30366303
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366303 250 mg 250mg
30363691 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30366303 Supplier Toronto Research Chemicals Supplier No. TRCC291710250MG
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 319425-66-6
Molecular Formula C22H26N2O5
Molecular Weight (g/mol) 398.45
Synonym CF 1743, 3-(2-Deoxy-β-D-erythro-pentofuranosyl)-6-(4-pentylphenyl)-furo[2,3-d]pyrimidin-2(3H)-one
IUPAC Name 3-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-(4-pentylphenyl)furo[2,3-d]pyrimidin-2-one
SMILES CCCCCc1ccc(cc1)C2=CC3=CN([C@H]4C[C@H](O)[C@@H](CO)O4)C(=O)N=C3O2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C22H26N2O5/c1-2-3-4-5-14-6-8-15(9-7-14)18-10-16-12-24(22(27)23-21(16)29-18)20-11-17(26)19(13-25)28-20/h6-10,12,17,19-20,25-26H,2-5,11,13H2,1H3/t17-,19+,20+/m0/s1
Formula Weight 398.1842
Chemical Name or Material 3-(2-Deoxy-beta-D-erythro-pentofuranosyl)-6-(4-pentylphenyl)-furo[2,3-d]pyrimidin-2(3H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.