missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios Dermanios Scrub CG

Product Code. 15195264
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15195264 500 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15195264 Supplier Laboratoires Anios Supplier No. 2082522

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antiseptic soap for hygienic handwashing.

Antiseptic Soap:

  • Clear solution, slightly viscous
  • Product pH : approx. 5
  • Density : approx. 1.0

Specifications

Product Type CG Dermanios Scrub
Composition Digluconate de chlorhexidine excipients
For Use With (Application) For Surgical Environment, Medical and Industrial Clean Room
Quantity 12 Pk.
Volume (Metric) 500 mL
compliance-icons
Product Identifier
  • DERMANIOS SCRUB CG
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P273-Avoid release to the environment.
  • P401-Store .
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: D-GLUCONIC ACID, COMPOUND WITH N, N''-BIS(4-CHLOROPHENYL)-3, 12- DIIMINO-2, 4,11, 13-TETRAAZATETRA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.