missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CGP 12177 hydrochloride

Product Code. 30001205
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001205 50 mg 50mg
18005472 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001205 Supplier R&D Systems™ Supplier No. 1134/50
  • ONLINE EXCLUSIVE PRICE
    420.50 €
    379.05 € / 50mg
    Save 41.45 €
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. β3 partial agonist. β1/β2 antagonist

Partial agonist at β3-adrenoceptors. β1- and β2-adrenoceptor antagonist (Ki values are 0.9, 4 and 88 nM for β1, β2 and β3 receptors respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64208-32-8
Molecular Formula C14H22ClN3O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 315.8
Purity >98%
Molecular Weight (g/mol) 315.798
Chemical Name or Material CGP 12177 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.