missing translation for 'onlineSavingsMsg'
Learn More

CGS 12066B dimaleate, 98+%, Thermo Scientific Chemicals

Product Code. 15485969
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485969 10 mg 10mg
15495969 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485969 Supplier Thermo Scientific Alfa Aesar Supplier No. J63801.MA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5-HT-1B full agonist

5-HT-1B full agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109028-10-6
Molecular Formula C25H25F3N4O8
Molecular Weight (g/mol) 566.49
MDL Number MFCD00055048
InChI Key HTEVMLYDEWVIQE-LVEZLNDCSA-N
Synonym cgs 12066b dimaleate, 1-methyl-4-7-trifluoromethyl pyrrolo 1,2-a quinoxalin-4-yl piperazine; bis butenedioic acid
PubChem CID 71299720
IUPAC Name (E)-but-2-enedioic acid;4-(4-methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline
SMILES CN1CCN(CC1)C2=NC3=C(C=CC(=C3)C(F)(F)F)N4C2=CC=C4.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O

Specifications

Melting Point 126°C to 127°C
Color White
Quantity 10 mg
Formula Weight 566.49
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material CGS 12066B dimaleate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.