missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress CH 5450

Product Code. 16443177
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16443177 Supplier MedChemExpress Supplier No. HY16707
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress CH 5450 (Z-Ile-Glu-Pro-Phe-Ome) is a human chymase inhibitor.

Specifications

Purity 99.47%
CAS 252557-97-4
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) COVID-19-immunoregulation
Molecular Weight (g/mol) 652.73 g/mol
Product Type Peptides
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 43 mg/mL (65.88 mM)
Synonym Z-Ile-Glu-Pro-Phe-Ome
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.