missing translation for 'onlineSavingsMsg'
Learn More

Melford CHAPS

Product Code. 30018161
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
1 kg
Unit Size:
10g
1g
1kg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018161 5 g 5g
30018157 1 g 1g
30018159 1 kg 1kg
30018158 10 g 10g
30018160 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30018161 Supplier Melford Supplier No. C410105.0
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Zwitterionic, non-denaturing detergent for solubilizing membrane proteins.

  • CAS Number: 75621-03-3
  • Molecular Formula: C32H58N2SO7
  • Molecular Weight: 614.9

Specifications

CAS 75621-03-3
Molecular Formula C32H58N2O7S
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.