missing translation for 'onlineSavingsMsg'
Learn More

CHAPS, MedChemExpress

Product Code. 16499097
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499097 5 g 5g
16461519 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499097 Supplier Medchem Express Supplier No. HY15435
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CHAPS, a derivative of Cholic acid, is a zwitterionic detergent for solubilizing membrane proteins. CHAPS is used for stabilization of various protein-DNA complexes and can retain biochemical activity of proteins in solution.

CHAPS, a derivative of Cholic acid, is a zwitterionic detergent for solubilizing membrane proteins. CHAPS is used for stabilization of various protein-DNA complexes and can retain biochemical activity of proteins in solution.

Specifications

Chemical Name or Material CHAPS
CAS 75621-03-3
Color White
Purity Grade Notes Research
Molecular Formula C32H58N2O7S
Quantity 5 g
Solubility Information H2O : 83.33 mg/mL (135.52 mM; ultrasonic and warming and heat to 60°C)
SMILES O[C@@H]1CC[C@]2(C)[C@@]3([H])C[C@H](O)[C@]4(C)[C@@H]([C@H](C)CCC(NCCC[N+](C)(C)CCCS([O-])(=O)=O)=O)CC[C@@]4([H])[C@]3([H])[C@H](O)C[C@@]([H])2C1
Molecular Weight (g/mol) 614.88
Formula Weight 614.88
Percent Purity 96.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.