missing translation for 'onlineSavingsMsg'
Learn More

CHAPS, MP Biomedicals™

Product Code. 11481632 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481632 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11481632 Supplier MP Biomedicals Supplier No. 04808196
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHAPS, ≥98%, White Powder

  • CHAPS is useful for solubilizing membrane proteins.
  • Frequently used prior to electrophoresis since it generally does not denature the proteins.
  • CHAPS is a Zwitterionic detergent.

Specifications

Color White
CAS 75621-03-3
Physical Form Powder
Melting Point 157°C (decomposition)(literature)
pH 4.0 to 7.0 (10% aq. soln.)
Quantity 5 g
Molecular Formula C32H58N2O7S
Synonym 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, CHAPS hydrate
Solubility Information Soluble in 10% aqueous solution.(Lit.)
InChI Key UMCMPZBLKLEWAF-BCTGSCMUSA-N
Molecular Weight (g/mol) 614.88
Percent Purity ≥98%
Grade Ungraded
Chemical Name or Material 3-(3-Cholamidopropyl)dimethylammonio-1-propanesulfonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.