missing translation for 'onlineSavingsMsg'
Learn More

CHAPS, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15578264 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15578264 100 g Poly Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15578264 Supplier Avantor J.T.Baker Supplier No. 414502

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Laboratory,Research,or Manufacturing Use

Specifications

CAS 75621-03-3
Packaging Poly Bottle
Molecular Formula C32H58N2O7S
Quantity 100 g
Specific Gravity 1L = 0.00kg
Synonym 3-(3-cholamidopropyl)dimethylammonio-1-propanesulfonate
InChI Key UMCMPZBLKLEWAF-GBSSQDIOSA-N
SMILES CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
Molecular Weight (g/mol) 614.883
PubChem CID 134129639
Formula Weight 614.89
Grade Ultrapure Bioreagent
Chemical Name or Material CHAPS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.