missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAPS (White Crystalline Powder), Fisher BioReagents™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
Description
CHAPS is a Zwitterionic detergent used for solubilizing membrane proteins.
Specifications
Absorbance | 0.1 max. (1%) at 260nm, 0.1 max. (1%) at 280nm |
Color | White |
Physical Form | Solid |
Melting Point | 267°C |
Assay Percent Range | ≥98 % |
Quantity | 5 g |
Merck Index | 15, 2043 |
Identification | Pass Test |
Formula Weight | 614.9 |
Packaging | Amber Glass |
Show More |
Safety and Handling
Product Identifier
- CHAPS
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'