missing translation for 'onlineSavingsMsg'
Learn More

CHAPS (White Crystalline Powder), Fisher BioReagents™

Product Code. 10659163
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10659163 1 g Amber Glass 1g
10274723 5 g Amber Glass 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10659163 Supplier Fisher BioReagents Supplier No. BP5711

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
CHAPS is a Zwitterionic detergent used for solubilizing membrane proteins.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75621-03-3
Molecular Formula C32H58N2O7S
Molecular Weight (g/mol) 614.883
InChI Key UMCMPZBLKLEWAF-GBSSQDIOSA-N
Synonym 3-[(3-Cholamidopropyl) dimethylammonio]-1-propanesulfonate
PubChem CID 134129639
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
SMILES CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Absorbance 0.1 max. (1%) at 260nm, 0.1 max. (1%) at 280nm
Color White
Physical Form Solid
Melting Point 267°C
Quantity 1 g
Identification Pass Test
Packaging Amber Glass
Assay Percent Range ≥98 %
Merck Index 15, 2043
Formula Weight 614.9
Chemical Name or Material CHAPS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.