missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CHAPSO (3-[(3-cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate)

Product Code. 30245336 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30245336 5 g 5g
30245085 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30245336 Supplier Thermo Scientific™ Supplier No. A65502
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHAPSO, or 3-[(3-cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate, is a zwitterionic detergent commonly used in biochemical and biophysical research.

CHAPSO, or 3-[(3-cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate, is a zwitterionic detergent commonly used in biochemical and biophysical research. It is widely employed for membrane protein solubilization, stabilization, and purification. CHAPSO is known for its mildness and ability to preserve the structural integrity and functionality of membrane proteins during extraction. Its unique properties make it suitable for a variety of applications, including protein crystallization, spectroscopy, and structural biology studies. With its versatile and effective solubilization capabilities, CHAPSO is a valuable tool for researchers working with membrane proteins.

Features of CHAPSO include:

  • Zwitterionic detergent
  • Gentle—enabling preservation of native conformation and activity
  • Effectively disrupts lipid-protein interactions
  • Exhibits low critical micelle concentration (CMC)
  • High-purity compound with low UV absorptivity

Properties

  • CMC: (H2O) ∼8 mM (0.50%)
  • Aggregation number: (H2O) ∼11
  • Purity: ≥99% (by HPLC analysis)
  • pH: 5–8 (1% solution in water)
  • Solubility: ≥0.5 M (in water at 20 °C)
  • Fluorescence: <10% (0.1% solution in water at 345 nm)
  • Absorbance of a 1% solution in water: 260 nm: <0.06, 280 nm: <0.04, 340 nm: <0.02
TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Detergent
CAS 82473-24-3
Molecular Formula C32H58N2O8S
Solubility ≥0.5 M (in water at 20°C)
Content And Storage Store below -20°C.
Form Lyophilized
Reagent Type Detergent
Purification Target Proteins
Quantity 5 g
Molecular Weight (g/mol) 630.9
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.