missing translation for 'onlineSavingsMsg'
Learn More

Charcoal, Activated, Powder, U.S.P., J.T. Baker™

Product Code. 15537914 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 15537914

Brand: Avantor J.T.Baker 156001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7440-44-0
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C
Quantity 500 g
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES [C]
IUPAC Name carbon
Molecular Weight (g/mol) 12.011
PubChem CID 5462310
ChEBI CHEBI:27594
Formula Weight 12.01
Grade USP
Physical Form Powder
Chemical Name or Material Charcoal, Activated
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.