missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free/Total

Product Code. 10769674
Change view
Click to view available options
Range:
0 to 1mg/L
0 to 3.5mg/L
0 to 4mg/L
0.02 to 0.3mg/L
0.1 to 2mg/L
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
10769674 0.1 to 2mg/L Each
10094454 0 to 3.5mg/L Each
11745458 0 to 4mg/L Each
10700255 0 to 4mg/L Each
11775458 0.1 to 2mg/L Each
11715458 0.02 to 0.3mg/L Each
10326622 0.02 to 0.3mg/L Each
11725458 0 to 1mg/L Each
10663713 0 to 1mg/L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10769674 Supplier Lovibond™ Supplier No. 147040
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Aqua Comparator Housing (D55 for L147000)
  • Aqua Comparator Disc
  • DPD No. 1 Rapid Tablets
  • DPD No. 3 Rapid Tablets
  • 2 x 13.5mm Sample Cells
  • Tablet Crusher
  • Instructions
  • Carrying case

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Accuracy ±5%
For Use With (Application) Routine water analysis
Includes Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
No. of Tests 100
Range 0.1 to 2mg/L
Type Chlorine Free and Total Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.