missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ CHECKIT™ Comparator Test Kits
Click to view available options
Range:
0 to 5mg/L
0-5.0mg/L
Type:
Bromine Color Disc
Copper
Unit Size:
Each
Description
Includes
Carrying case, comparator, color disc, sample cells, instructions and enough reagents for 100 tests
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
- Color discs and reagents are available separately
Specifications
Specifications
| For Use With (Application) | For routine water analysis |
| For Use With (Equipment) | Aqua comparator™ |
| Range | 0 to 5mg/L |
| Sufficient For | 100 Tests |
| Type | Bromine Color Disc |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction