missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Checkit™ Comparators 2-in-1 Test Kits

Product Code. 10193202
Change view
Click to view available options
Range:
6.5 to 8.4 pH, 0 to 1 mg/L Cl2
6.5 to 8.4 pH, 0 to 1 mg/L Cu
6.5 to 8.4 pH, 0 to 4 mg/L Cl2
6.5 to 8.4 pH, 0 to 5 mg/L Br
6.5 to 8.4 pH, 0.1 to 2 mg/L Cl2
Description:
pH Value
Copper
pH Value
Bromine
pH Value
Chlorine
pH value
Chlorine
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Description unitSize
10193202 6.5 to 8.4 pH, 0 to 1 mg/L Cl2 pH Value
Chlorine
Each
10631613 6.5 to 8.4 pH, 0.1 to 2 mg/L Cl2 pH Value
Chlorine
Each
10739355 6.5 to 8.4 pH, 0 to 1 mg/L Cu pH Value
Copper
Each
11735458 6.5 to 8.4 pH, 0 to 1 mg/L Cl2 pH Value
Chlorine
Each
10739104 6.5 to 8.4 pH, 0 to 4 mg/L Cl2 pH value
Chlorine
Each
10461045 6.5 to 8.4 pH, 0 to 5 mg/L Br pH Value
Bromine
Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10193202 Supplier Lovibond™ Supplier No. 147016
  • ONLINE EXCLUSIVE PRICE
    101.00 €
    99.70 € / Pack of 1
    Save 1.30 €
    1% Off
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.

  • Easy operation and exact reagent dosing
  • Tablet reagents with a minimm guaranteed shelf life of 5/10 years
  • Rotate continuous gradient color wheel to obtain a color match with the reacted sample
  • Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks

Specifications

Description pH Value
Chlorine
Includes Checkit Discs, cells, stirring rod and Aqualytic tablet reagents (for 30 Tests) for the desired test
No. of Tests 30
Range 6.5 to 8.4 pH, 0 to 1 mg/L Cl2
Sufficient For 30 Tests
Test Method Colorimetric
Type 2 in 1 Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.