missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics Mercury-Free COD Vials Kits

Product Code. 11761044
Change view
Click to view available options
Range:
0 to 15,000 ppm
0 to 15,000 ppm (HR+)
0 to 150 ppm (LR)
0 to 1500 ppm
Includes:
150 vials and instruction book
25 vials and instruction book
98 vials and instruction book
No. of Tests:
149 (except distilled water)
24
97 (except distilled water)
Unit Size:
0.01 L
7.13 L
Pack of 25
Pack of 98
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Includes No. of Tests Unit Size
11761044 0 to 1500 ppm 150 vials and instruction book 149 (except distilled water) 7.13 L
11893510 0 to 150 ppm (LR) 25 vials and instruction book 24 Pack of 25
11813520 0 to 15,000 ppm (HR+) 25 vials and instruction book 24 0.01 L
11791054 0 to 15,000 ppm 98 vials and instruction book 97 (except distilled water) Pack of 98
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11761044 Supplier CHEMetrics Supplier No. K7366
  • ONLINE EXCLUSIVE PRICE
    237.81 €
    196.01 € / 7.13 L
    Save 41.80 €
    17% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Contains premeasured reagents for accurate, economical determination of COD

  • No need to modify or recalibrate existing equipment or procedures
  • Optical-quality glass for accuracy
  • Available in mercury and mercury-free versions
  • Standard 5/8 in. (16 mm) diameter for compatibility with standard COD reactors and spectrophotometers; can also be used in micro-COD reactors
  • Follow the EPA-approved method for compliance reporting and use potassium dichromate as the oxidizing agent, silver sulfate as the catalyst, and mercury chloride to remove chloride interference
  • Mercury-free version uses the same oxidizing agent and catalyst without the chloride compensator

Specifications

Analytes Chemical Oxygen Demand (COD)
Description A fresh reagent ampoule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.
For Use With (Application) Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater
For Use With (Equipment) Digestor block as well as a photometer or spectrophotometer
Includes 150 vials and instruction book
No. of Tests 149 (except distilled water)
Range 0 to 1500 ppm
Test Method Dichromate reactor digestion
Test Time 2 hr. digestion
Type COD Vials Kit
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • COD Vials Kit 0-1500 ppm (HR) Mercury-free
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P363-Wash contaminated clothing before reuse.
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.