missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics Mercury-Free COD Vials Kits

Contains premeasured reagents for accurate, economical determination of COD

Brand:  CHEMetrics K7361S

Code : A0

Additional Details : CAS Number : 7664-93-9 Weight : 0.51750kg Transport : UN number : 1830 Chem class : 8 Pack group : II


 View more versions of this product

Product Code. 11803520

  • 38.60 € / 0.08 L
Estimated Shipment: 29-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • No need to modify or recalibrate existing equipment or procedures
  • Optical-quality glass for accuracy
  • Available in mercury and mercury-free versions
  • Standard 5/8 in. (16 mm) diameter for compatibility with standard COD reactors and spectrophotometers; can also be used in micro-COD reactors
  • Follow the EPA-approved method for compliance reporting and use potassium dichromate as the oxidizing agent, silver sulfate as the catalyst, and mercury chloride to remove chloride interference
  • Mercury-free version uses the same oxidizing agent and catalyst without the chloride compensator

Specifications

Specifications

Chemical Oxygen Demand (COD)
Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater
25 vials and instruction book
0 to 1500 ppm (HR)
2 hr. digestion
A fresh reagent ampoule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.
Digestor block as well as a photometer or spectrophotometer
24
Dichromate Reactor Digestion
COD Vials Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • COD Reagent Vials 0-1500 ppm (HR) Mercury-free

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 1B
  • Oxidising liquid Category 3
  • Reproductive toxicity Category 1A
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers