Learn More
Medimark International™ Chemgene™ HLD4L Laboratory Disinfectant
For multipurpose surface disinfection and cleaning. Chemgene™ HLD4L Lab Disinfectant destroys bacteria, fungi, mycobacteria, viruses and spores.
Brand:  Medimark International™ XTM305
Code : 04
Additional Details : CAS Number : 68424-85-1 Weight : 0.98100kg Transport : UN number : 1760 Chem class : 8 Pack group : III
 View more versions of this product
Product Code. 15136686
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
For multipurpose surface disinfection and cleaning. Chemgene™ Lab Disinfectant Spray destroys bacteria, fungi, mycobacteria, viruses and spores.
Chemgene HLD4L disinfectant concentrate.
  • Manufactured in accordance with Quality Management System ISO 9001
  • Biodegradable under OECD conditions
  • Denatures DNA/RNA
Specifications
Specifications
Lab Disinfectant
0.048611111111
1 L
Clear
For multipurpose Surface disinfection and cleaning
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • CHEMGENE HLD4L CONCENTRATE
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P315-Get immediate medical advice/attention.